Benzyl (S)-(+)-glycidyl ether 97%

Supplier: Apollo Scientific

Synonyms: (S)-2-((Benzyloxy)methyl)oxirane

OR315269-25G
APOSOR315269-25GEA 69.8 EUR
APOSOR315269-25G
Benzyl (S)-(+)-glycidyl ether 97%
Benzyl (S)-(+)-glycidyl ether
Formula: C₁₀H₁₂O₂
MW: 164,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00054428
CAS Number: 16495-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR