4-(Benzyloxy)-2-hydroxybenzaldehyde 97%

Supplier: Apollo Scientific

APOSOR315520-5GEA 85.8 EUR
APOSOR315520-5G
4-(Benzyloxy)-2-hydroxybenzaldehyde 97%
4-(Benzyloxy)-2-hydroxybenzaldehyde
Formula: C₁₄H₁₂O₃
MW: 228,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01075698
CAS Number: 52085-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR