2-Cyclopropylbenzonitrile 95+%

Supplier: Apollo Scientific

APOSOR318044-10GEA 2000 EUR
APOSOR318044-10G APOSOR318044-1G APOSOR318044-5G
2-Cyclopropylbenzonitrile 95+%
2-Cyclopropylbenzonitrile
Formula: C₁₀H₁₀N
Storage Temperature: Ambient
MDL Number: MFCD00463731
CAS Number: 3154-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR