Cyclopropanesulphonamide 98%
Supplier: Apollo Scientific
|
Warning
|
APOSOR3194-1GEA
14.7
EUR
APOSOR3194-1G
Cyclopropanesulphonamide 98%
Cyclopropanesulphonamide
Formula:
C₃H₇NO₂S MW: 121,15 g/mol Melting Pt: 101…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD08705286 CAS Number: 154350-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...