3-Methyl-2-butyl isothiocyanate

Supplier: Apollo Scientific

APOSOR322141-1GEA 139 EUR
APOSOR322141-1G
3-Methyl-2-butyl isothiocyanate
3-Methyl-2-butyl isothiocyanate
Formula: C₆H₁₁NS
Storage Temperature: Ambient
MDL Number: MFCD00060388
CAS Number: 201224-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR