1-(2-Pyrimidyl)piperazine dihydrochloride

Synonyms: 2-(1-Piperazinyl)pyrimidine Dihydrochloride

APOSOR322273-5GEA 0 EUR
APOSOR322273-5G
1-(2-Pyrimidyl)piperazine dihydrochloride
1-(2-Pyrimidyl)piperazine dihydrochloride
Formula: C₈H₁₂N₄
MW: 237,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012779
CAS Number: 94021-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR