(S)-(+)-2-Hexyl isothiocyanate
APOSOR322564-1GEA
0
EUR
APOSOR322564-1G
(S)-(+)-2-Hexyl isothiocyanate
(S)-(+)-2-Hexyl isothiocyanate
Formula:
C₇H₁₃NS MW: 143,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05664054 CAS Number: 737000-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...