3-Iodo-1H-pyrazole
Supplier: Apollo Scientific
Synonyms:
3-Iodopyrazole
APOSOR322575-1GEA
53.9
EUR
APOSOR322575-1G
3-Iodo-1H-pyrazole
3-Iodo-1H-pyrazole
Formula:
C₃H₃IN₂ MW: 193,98 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12022324 CAS Number: 4522-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...