(R)-(-)-2-Heptyl isothiocyanate

Supplier: Apollo Scientific

APOSOR322587-1GEA 196 EUR
APOSOR322587-1G
(R)-(-)-2-Heptyl isothiocyanate
(R)-(-)-2-Heptyl isothiocyanate
Formula: C₈H₁₅NS
Storage Temperature: Ambient
MDL Number: MFCD05664051
CAS Number: 737000-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR