1,4-Bis(aminomethyl)cyclohexane (cis and trans mixture)

Synonyms: Hexahydro-p-xylylenediamine1,4-Di(aminomethyl)cyclohexane

APOSOR322591-25GEA 0 EUR
APOSOR322591-25G
1,4-Bis(aminomethyl)cyclohexane (cis and trans mixture)
1,4-Bis(aminomethyl)cyclohexane (cis and trans mixture)
Formula: C₈H₁₈N₂
MW: 142,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039463
CAS Number: 2549-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR