1-(6-Chloro-3-pyridazinyl)-4-piperidinecarboxamide

Supplier: Apollo Scientific

Synonyms: 1-(6-Chloro-3-pyridazinyl)piperidine-4-carboxamide

APOSOR322667-500MGEA 285 EUR
APOSOR322667-500MG
1-(6-Chloro-3-pyridazinyl)-4-piperidinecarboxamide
1-(6-Chloro-3-pyridazinyl)-4-piperidinecarboxamide
Formula: C₁₀H₁₃ClN₄O
MW: 240,69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00180782
CAS Number: 303149-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR