3-(4-Methylphenyl)-1H-pyrazol-5-amine

Supplier: Apollo Scientific
Warning

Synonyms: 5-Amino-3-(4-methylphenyl)pyrazole

OR3416-5G
APOSOR3416-5GEA 219 EUR
APOSOR3416-5G
3-(4-Methylphenyl)-1H-pyrazol-5-amine
3-(4-Methylphenyl)-1H-pyrazol-5-amine
Formula: C₁₀H₁₁N₃
Melting Pt: 148…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00084887
CAS Number: 78597-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR