5-Acetyl-2-methylpyridine
Supplier: Apollo Scientific
|
Warning
|
APOSOR345083-1GEA
299
EUR
APOSOR345083-1G
5-Acetyl-2-methylpyridine
5-Acetyl-2-methylpyridine
Formula:
C₈H₉NO MW: 135,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00128113 CAS Number: 36357-38-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...