2-Amino-5-bromo-3-methoxypyridine 98%
|
Warning
|
Synonyms:
5-Bromo-3-methoxypyridin-2-amine
APOSOR345479-10GEA
0
EUR
APOSOR345479-10G
APOSOR345479-1G
APOSOR345479-5G
2-Amino-5-bromo-3-methoxypyridine 98%
2-Amino-5-bromo-3-methoxypyridine
Formula:
C₆H₇BrN₂O MW: 203,04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09909849 CAS Number: 42409-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...