4-(2-Bromophenylcarbamoyl)phenylboronic acid 95%

Supplier: Apollo Scientific
Warning

Synonyms: (4-((2-Bromophenyl)carbamoyl)phenyl)boronic acid

APOSOR3498-1GEA 14.7 EUR
APOSOR3498-1G
4-(2-Bromophenylcarbamoyl)phenylboronic acid 95%
4-(2-Bromophenylcarbamoyl)phenylboronic acid
Formula: C₁₃H₁₁BBrNO₃
MW: 319,9500 g/mol
Melting Pt: 233…235 °C
Storage Temperature: Ambient
MDL Number: MFCD09027202
CAS Number: 874288-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR