4-(Dimethylamino)cinnamic acid

Supplier: Apollo Scientific

APOSOR350131-5GEA 141 EUR
APOSOR350131-5G
4-(Dimethylamino)cinnamic acid
4-(Dimethylamino)cinnamic acid
Formula: C₁₁H₁₃NO₂
MW: 191,23
Storage Temperature: Ambient
MDL Number: MFCD00004397
CAS Number: 1552-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR