2-(4-Methyl-5-thiazolyl)ethyl acetate
Synonyms:
4-Methyl-5-thiazolylethyl acetate
, Acetic Acid 2-(4-Methyl-5-thiazolyl)ethyl Ester
, 5-(2-Acetoxyethyl)-4-methylthiazole
APOSOR350238-100GEA
0
EUR
APOSOR350238-100G
2-(4-Methyl-5-thiazolyl)ethyl acetate
2-(4-Methyl-5-thiazolyl)ethyl acetate
Formula:
C₈H₁₁NO₂S MW: 185,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005338 CAS Number: 656-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...