4-(Methylsulphinyl)phenol 98%

APOSOR3525-5GEA 0 EUR
APOSOR3525-5G APOSOR3525-1G
4-(Methylsulphinyl)phenol 98%
4-(Methylsulphinyl)phenol
Formula: C₇H₈O₂S
Melting Pt: 46…50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00464311
CAS Number: 14763-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR