1-(Phenylsulphonyl)-5-indoleboronic acid

Supplier: Apollo Scientific

APOSOR3696-1GEA 218 EUR
APOSOR3696-1G
1-(Phenylsulphonyl)-5-indoleboronic acid
1-(Phenylsulphonyl)-5-indoleboronic acid
Formula: C₁₄H₁₂BNO₄S
Storage Temperature: Ambient
MDL Number: MFCD03840606
CAS Number: 480438-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR