4-(Methylsulphonyl)phenylboronic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-(Methanesulphonyl)phenylboronic acid
, 4-(Methylsulphonyl)benzeneboronic acid
APOSOR3734-1GEA
14.7
EUR
APOSOR3734-1G
APOSOR3734-5G
4-(Methylsulphonyl)phenylboronic acid
4-(Methylsulphonyl)phenylboronic acid
Formula:
C₇H₉BO₄S MW: 200,02 g/mol Melting Pt: 289…293 °C Storage Temperature: Ambient |
MDL Number:
MFCD01630820 CAS Number: 149104-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...