4-sec-Butylphenylboronic acid
Synonyms:
4-sec-Butylbenzeneboronic acid
APOSOR3941-1GEA
154
EUR
APOSOR3941-1G
APOSOR3941-10G
APOSOR3941-5G
4-sec-Butylphenylboronic acid
4-sec-Butylphenylboronic acid
Formula:
C₁₀H₁₅BO₂ MW: 178,04 g/mol Melting Pt: 96…100 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02179456 CAS Number: 850568-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...