3-(Ethylcarbamoyl)phenylboronic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: 3-(Ethylcarbamoyl)benzeneboronic acid

APOSOR3954-1GEA 136 EUR
APOSOR3954-1G APOSOR3954-5G
3-(Ethylcarbamoyl)phenylboronic acid 97%
3-(Ethylcarbamoyl)phenylboronic acid
Formula: C₉H₁₂BNO₃
MW: 193,01
Melting Pt: 106…116 °C
Storage Temperature: Ambient
MDL Number: MFCD04115689
CAS Number: 850567-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR