4-Bromo-3-methyl-1H-indazole 95+%

Supplier: Apollo Scientific

APOSOR40117-250MGEA 74.2 EUR
APOSOR40117-250MG
4-Bromo-3-methyl-1H-indazole 95+%
4-Bromo-3-methyl-1H-indazole
Formula: C₈H₇BrN₂
MW: 211,0586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743499
CAS Number: 1159511-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR