3-(4-Chlorophenyl)-1H-pyrazol-5-amine

Supplier: Apollo Scientific
Danger

Synonyms: 5-Amino-3-(4-chlorophenyl)-1H-pyrazole

APOSOR4080-1GEA 50.1 EUR
APOSOR4080-1G APOSOR4080-25G APOSOR4080-5G
3-(4-Chlorophenyl)-1H-pyrazol-5-amine
3-(4-Chlorophenyl)-1H-pyrazol-5-amine
Formula: C₉H₈ClN₃
MW: 193,64
Melting Pt: 171…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00053046
CAS Number: 78583-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR