2-Piperazinonicotinonitrile 98%
Supplier: Apollo Scientific
|
Danger
|
OR4195-250MG
APOSOR4195-250MGEA
17.6
EUR
APOSOR4195-250MG
2-Piperazinonicotinonitrile 98%
2-Piperazinonicotinonitrile
Formula:
C₁₀H₁₂N₄ Melting Pt: 76…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190248 CAS Number: 84951-44-0 UN: 2923 ADR: 8+6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...