(2-Benzimidazolyl)acetonitrile 98%
Supplier: Apollo Scientific
APOSOR42023-25GEA
72.7
EUR
APOSOR42023-25G
APOSOR42023-100G
(2-Benzimidazolyl)acetonitrile 98%
(2-Benzimidazolyl)acetonitrile
Formula:
C₉H₇N₃ MW: 157,18 g/mol Melting Pt: 200…205 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005601 CAS Number: 4414-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...