N-Benzyl-3-borono-4-methylbenzenesulphonamide 97%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Benzylsulfamoyl-2-methylbenzeneboronic acid

APOSOR4234-250MGEA 123 EUR
APOSOR4234-1G APOSOR4234-500MG APOSOR4234-250MG
N-Benzyl-3-borono-4-methylbenzenesulphonamide 97%
N-Benzyl 3-borono-4-methylbenzenesulphonamide
Formula: C₁₄H₁₆BNO₄S
MW: 305,16
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD07363760
CAS Number: 871329-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR