3-Amino-5-phenyl-1H-pyrazole 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Amino-5-phenylpyrazole
APOSOR4440-5GEA
23.4
EUR
APOSOR4440-5G
APOSOR4440-10G
3-Amino-5-phenyl-1H-pyrazole 97%
3-Amino-5-phenyl-1H-pyrazole
Formula:
C₉H₉N₃ MW: 159,19 g/mol Melting Pt: 118…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191749 CAS Number: 1572-10-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...