4-Benzyloxybenzophenone 98+%
Supplier: Apollo Scientific
|
Warning
|
APOSOR451318-5GEA
21.9
EUR
APOSOR451318-5G
APOSOR451318-25G
APOSOR451318-100G
4-Benzyloxybenzophenone 98+%
4-Benzyloxybenzophenone
Formula:
C₁₄H₉F₃O Melting Pt: 82…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017508 CAS Number: 54589-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...