Citric acid monohydrate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Hydroxytricarballylic acid
, 2-Hydroxy-1,2,3-propanetricarboxilic acid
APOSOR460066-1KGEA
109
EUR
APOSOR460066-1KG
APOSOR460066-2.5KG
Citric acid monohydrate
Citric acid monohydrate
Formula:
C₆H₈O₇.H₂O MW: 210,14 g/mol Melting Pt: 135…158 °C Density: 1,542 g/cm³ (25 °C) Flash Pt: 173,9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149972 CAS Number: 5949-29-1 EINECS: 201-069-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...