(S)-tert-Butyl-2-cyanopyrrolidine-1-carboxylate 97%

Supplier: Apollo Scientific

Synonyms: (S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinecarbonitrile(S)-1-Boc-2-cyanopyrrolidinetert-Butyl (S)-2-Cyano-1-pyrrolidinecarboxylate(S)-1-(tert-Butoxycarbonyl)-2-cyanopyrrolidine(S)-2-Cyano-1-pyrrolidinecarboxyic acid tert-Butyl ester(S)-1-Boc-2-pyrrolidinecarbonitrile

APOSOR4610-10GEA 71.4 EUR
APOSOR4610-10G
(S)-tert-Butyl-2-cyanopyrrolidine-1-carboxylate 97%
(S)-tert-Butyl-2-cyanopyrrolidine-1-carboxylate
Formula: C₁₀H₁₆N₂O₂
MW: 196,25
Storage Temperature: Ambient
MDL Number: MFCD01861224
CAS Number: 228244-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR