(S)-2-Aminobutanamide hydrochloride 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
H-Abu-NH2 · HCl
, H-Abu-NH2 HCl
APOSOR46608-100GEA
139
EUR
APOSOR46608-100G
(S)-2-Aminobutanamide hydrochloride 97%
(S)-2-Aminobutanamide hydrochloride
Formula:
C₄H₁₁ClN₂O MW: 138,60 g/mol Melting Pt: 259…263 °C Storage Temperature: Ambient |
MDL Number:
MFCD00136565 CAS Number: 7682-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...