Tricyclazole
Synonyms:
5-Methyl[1,2,4]triazolo[3,4-b][1,3]benzothiazole
APOSOR46739-1GEA
201
EUR
APOSOR46739-1G
Tricyclazole
Tricyclazole
Formula:
C₉H₇N₃S Storage Temperature: Ambient |
CAS Number:
41814-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...