1,2:7,8-Diepoxyoctane

Supplier: Apollo Scientific
Danger

Synonyms: 1,4-Di(oxiran-2-yl)butane

APOSOR471095-25GEA 154 EUR
APOSOR471095-25G
1,2:7,8-Diepoxyoctane
1,2:7,8-Diepoxyoctane
Formula: C₈H₁₄O₂
MW: 142,20 g/mol
Boiling Pt: 240 °C
Density: 0,997 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00005155
CAS Number: 2426-07-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR