6-Chloro-1H-indazole 98%
Supplier: Apollo Scientific
APOSOR471640-5GEA
118
EUR
APOSOR471640-5G
APOSOR471640-10G
6-Chloro-1H-indazole 98%
6-Chloro-1H-indazole
Formula:
C₇H₅ClN₂ MW: 152,5800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07774234 CAS Number: 698-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...