5-Amino-2-mercaptobenzimidazole 96%
Supplier: Apollo Scientific
Synonyms:
5-Amino-1H-benzoimidazole-2-thiol
APOSOR480589-100GEA
271
EUR
APOSOR480589-100G
5-Amino-2-mercaptobenzimidazole 96%
5-Amino-2-mercaptobenzimidazole
Formula:
C₇H₇N₃S MW: 165,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00022671 CAS Number: 2818-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...