4-(Carboxymethyl)phenylboronic acid

Supplier: Apollo Scientific
Warning

APOSOR48251-1GEA 382 EUR
APOSOR48251-1G APOSOR48251-250MG
4-(Carboxymethyl)phenylboronic acid
4-(Carboxymethyl)phenylboronic acid
Formula: C₈H₉BO₄
Storage Temperature: Refrigerator
MDL Number: MFCD03425919
CAS Number: 90111-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR