2-(Tributylstannyl)oxazole

Danger

APOSOR52460-1GEA 285 EUR
APOSOR52460-1G
2-(Tributylstannyl)oxazole
2-(Tributylstannyl)oxazole
Formula: C₁₅H₂₉NOSn
MW: 358,11 g/mol
Boiling Pt: 108 °C
Density: 1,71 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD06798076
CAS Number: 145214-05-7
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR