4-Ethylbenzylamine
Supplier: Apollo Scientific
|
Danger
|
APOSOR52606-5GEA
292
EUR
APOSOR52606-5G
4-Ethylbenzylamine
4-Ethylbenzylamine
Formula:
C₉H₁₃N Boiling Pt: 208 °C Storage Temperature: Ambient |
MDL Number:
MFCD01529871 CAS Number: 7441-43-2 UN: 2735 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...