Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride 95+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
, Methyl 1-Benzyl-4-oxo-3-piperidinecarboxylate hydrochloride
, 1-Benzyl-4-oxo-3-piperidinecarboxylic acid methyll ester hydrochloride
, 1-Benzyl-3-methoxycarbonyl-4-piperidone hydrochloride
APOSOR54576-100GEA
105
EUR
APOSOR54576-100G
Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride 95+%
Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
Formula:
C₁₄H₁₈ClNO₃ C₁₅H₁₉NO₃·HCl MW: 283,75 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00012799 CAS Number: 3939-01-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...