2-Amino-2'-chloro-5-nitrobenzophenone

Supplier: Apollo Scientific
Warning

Synonyms: (2-Amino-5-nitrophenyl)(2-chlorophenyl)methanone

OR5767-5G
APOSOR5767-5GEA 14.7 EUR
APOSOR5767-5G
2-Amino-2'-chloro-5-nitrobenzophenone
2-Amino-2'-chloro-5-nitrobenzophenone
Formula: C₁₃H₉ClN₂O₃
MW: 276,68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00792453
CAS Number: 2011-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR