3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid 97%
Supplier: Apollo Scientific
Synonyms:
(3-Chloro-4-(cyclopropylcarbamoyl)phenyl)boronic acid
, 3-Chloro-4-(cyclopropylcarbamoyl)benzeneboronic acid
APOSOR5816-1GEA
193
EUR
APOSOR5816-1G
3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid 97%
3-Chloro-4-(cyclopropylcarbamoyl)phenylboronic acid
Formula:
C₁₀H₁₁BClNO₃ MW: 239,46 g/mol Melting Pt: 216…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD07363770 CAS Number: 850589-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...