1H-Indazole-6-carboxylic acid 95%

Supplier: Apollo Scientific
Warning

APOSOR60063-1GEA 18.8 EUR
APOSOR60063-1G APOSOR60063-250MG APOSOR60063-5G
1H-Indazole-6-carboxylic acid 95%
1H-Indazole-6-carboxylic acid
Formula: C₈H₆N₂O₂
Melting Pt: 297…300 °C
Storage Temperature: Ambient
MDL Number: MFCD06804571
CAS Number: 704-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR