2'-O-Methyluridine 98+%

Supplier: Apollo Scientific
Warning

APOSOR60199-5GEA 62.6 EUR
APOSOR60199-25G APOSOR60199-1G APOSOR60199-5G
2'-O-Methyluridine 98+%
2'-O-Methyluridine
Formula: C₁₀H₁₄N₂O₆
Melting Pt: 157…163 °C
Storage Temperature: Freezer
MDL Number: MFCD00056054
CAS Number: 2140-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR