2-Amino-3-pyridinemethanol

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-3-hydroxymethylpyridine2-Amino-3-(hydroxymethyl)pyridine

APOSOR6117-1GEA 24.8 EUR
APOSOR6117-1G
2-Amino-3-pyridinemethanol
2-Amino-3-pyridinemethanol
Formula: C₆H₈N₂O
MW: 124,14 g/mol
Boiling Pt: 279 °C
Melting Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD05663510
CAS Number: 23612-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR