Benzo[b]thiophene-2-carboxaldehyde 96%

Supplier: Apollo Scientific
Warning

APOSOR8022-1GEA 77.7 EUR
APOSOR8022-1G APOSOR8022-500MG
Benzo[b]thiophene-2-carboxaldehyde 96%
Benzo[b]thiophene-2-carboxaldehyde
Formula: C₉H₆OS
MW: 162,21
Boiling Pt: 90 °C/0,1mmHg
Storage Temperature: Ambient
MDL Number: MFCD01075041
CAS Number: 3541-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR