4-Ethoxybenzylamine 98%
Supplier: Apollo Scientific
Synonyms:
(4-Ethoxyphenyl)methanamine
APOSOR909510-25GEA
328
EUR
APOSOR909510-25G
APOSOR909510-100G
APOSOR909510-5G
4-Ethoxybenzylamine 98%
4-Ethoxybenzylamine
Formula:
C₉H₁₃NO MW: 151,2056 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02225920 CAS Number: 6850-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...