cis-3-Hexenyl acetate 97%
Supplier: Apollo Scientific
APOSOR916315-25GEA
147
EUR
APOSOR916315-25G
cis-3-Hexenyl acetate 97%
cis-3-Hexenyl acetate
Formula:
C₈H₁₄O₂ MW: 142,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00036563 CAS Number: 3681-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...