3-(2-Hydroxyethoxy)benzaldehyde 96%
Supplier: Apollo Scientific
OR924646-25G
OR924646-5G
APOSOR924646-25GEA
285
EUR
APOSOR924646-25G
APOSOR924646-5G
3-(2-Hydroxyethoxy)benzaldehyde 96%
3-(2-Hydroxyethoxy)benzaldehyde
Formula:
C₉H₁₀O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00191448 CAS Number: 60345-97-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...