3-Cyano-6-methylchromone 95%

Supplier: Apollo Scientific

Synonyms: 6-Methylchromone-3-carbonitrile

APOSOR925911-1GEA 113 EUR
APOSOR925911-1G APOSOR925911-5G
3-Cyano-6-methylchromone 95%
3-Cyano-6-methylchromone
Formula: C₁₁H₇NO₂
MW: 185,19
Storage Temperature: Ambient
MDL Number: MFCD00191962
CAS Number: 50743-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR